Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGCCAGGCTTCATGGATGAGCG[C/G]CGGCAGTTGGGACAGCTCCGGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601592 | ||||||||||||||||||||
Literature Links: |
RAPSN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RAPSN - receptor associated protein of the synapse | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005055.4 | 1423 | Silent Mutation | CGC,CGG | R,R 403 | NP_005046.2 | |
NM_032645.4 | 1423 | Silent Mutation | CGC,CGG | R,R 344 | NP_116034.2 | |
XM_005253042.3 | 1423 | Silent Mutation | CGC,CGG | R,R 385 | XP_005253099.1 | |
XM_005253043.3 | 1423 | Silent Mutation | CGC,CGG | R,R 362 | XP_005253100.1 | |
XM_011520252.1 | 1423 | Missense Mutation | CCG,GCG | P,A 432 | XP_011518554.1 |