Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCCAAATGAGCAATTTTGGTCA[A/G]CAAAGGGGTTATTTAGCTCCTAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600925 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRJ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRJ - protein tyrosine phosphatase, receptor type J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098503.1 | Intron | NP_001091973.1 | ||||
NM_002843.3 | Intron | NP_002834.3 | ||||
XM_017018083.1 | Intron | XP_016873572.1 | ||||
XM_017018084.1 | Intron | XP_016873573.1 | ||||
XM_017018085.1 | Intron | XP_016873574.1 |