Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTGTTTTGTTTCTGACAGACGC[A/T]AACGCACTCTTCTAGTAATGACTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1551 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1551 - KIAA1551 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018169.3 | Intron | NP_060639.3 | ||||
XM_005253405.2 | Intron | XP_005253462.1 | ||||
XM_006719099.2 | Intron | XP_006719162.1 | ||||
XM_011520722.1 | Intron | XP_011519024.1 | ||||
XM_017019550.1 | Intron | XP_016875039.1 |