Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCATTTTCCTACACCACTGACCC[A/G]AATATGCCCGGCGCCATGGGGACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613541 MIM: 602198 MIM: 605501 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf65 - chromosome 12 open reading frame 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143905.2 | 307 | Missense Mutation | CAA,CGA | Q,R 15 | NP_001137377.1 | |
NM_001194995.1 | 307 | Missense Mutation | CAA,CGA | Q,R 15 | NP_001181924.1 | |
NM_152269.4 | 307 | Missense Mutation | CAA,CGA | Q,R 15 | NP_689482.1 | |
XM_005253630.4 | 307 | Missense Mutation | CAA,CGA | Q,R 15 | XP_005253687.1 | |
XM_011538980.2 | 307 | Missense Mutation | CAA,CGA | Q,R 15 | XP_011537282.1 |
CDK2AP1 - cyclin dependent kinase 2 associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPHOSPH9 - M-phase phosphoprotein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |