Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTTGGGGGAGTCGGCAGCCGCCC[T/C]GCTCCTGCAGGGCCGTTCCTGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615927 MIM: 600848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM101A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM101A - family with sequence similarity 101 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCOR2 - nuclear receptor corepressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077261.3 | 7767 | UTR 3 | NP_001070729.2 | |||
NM_001206654.1 | 7767 | UTR 3 | NP_001193583.1 | |||
NM_006312.5 | 7767 | UTR 3 | NP_006303.4 |
ZNF664-FAM101A - filamin-interacting protein FAM101A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |