Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTTGTATGTTTTGTCCATGTTGT[A/G]TAGAGAACCACAGACTATTTCAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616245 MIM: 604878 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMC4 - ER membrane protein complex subunit 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A6 - solute carrier family 12 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042494.1 | 4396 | UTR 3 | NP_001035959.1 | |||
NM_001042495.1 | 4396 | UTR 3 | NP_001035960.1 | |||
NM_001042496.1 | 4396 | UTR 3 | NP_001035961.1 | |||
NM_001042497.1 | 4396 | UTR 3 | NP_001035962.1 | |||
NM_005135.2 | 4396 | UTR 3 | NP_005126.1 | |||
NM_133647.1 | 4396 | UTR 3 | NP_598408.1 | |||
XM_006720793.3 | 4396 | UTR 3 | XP_006720856.1 | |||
XM_011522269.2 | 4396 | Intron | XP_011520571.1 |