Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCCAGCCGGAGCTACCTGCT[C/T]AGCGTGCGGCCCGAGACCAGGTAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALPK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALPK3 - alpha kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020778.4 | 896 | Silent Mutation | CTC,CTT | L,L 243 | NP_065829.3 | |
XM_017022444.1 | 896 | Silent Mutation | CTC,CTT | L,L 243 | XP_016877933.1 | |
XM_017022445.1 | 896 | Silent Mutation | CTC,CTT | L,L 243 | XP_016877934.1 |