Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAACAGAATTGGAAATCCAAGTGG[C/T]AAATGGCAGATAAGGGTCCTGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHISA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SHISA9 - shisa family member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145204.2 | 1436 | Intron | NP_001138676.2 | |||
NM_001145205.1 | 1436 | UTR 3 | NP_001138677.2 | |||
XM_005255539.3 | 1436 | Intron | XP_005255596.2 | |||
XM_011522642.2 | 1436 | Intron | XP_011520944.1 |