Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGCTGTTGCCAGTCAAGTGAGG[A/C]TGAAGTTGAAATTCTGGGACCTTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SYT17 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SYT17 - synaptotagmin 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308157.1 | 550 | Missense Mutation | GAT,GCT | D,A 41 | NP_001295086.1 | |
NM_016524.3 | 550 | Missense Mutation | GAT,GCT | D,A 45 | NP_057608.2 | |
XM_005255362.3 | 550 | UTR 5 | XP_005255419.1 | |||
XM_011545865.1 | 550 | Missense Mutation | GAT,GCT | D,A 61 | XP_011544167.1 | |
XM_011545866.2 | 550 | Missense Mutation | GAT,GCT | D,A 52 | XP_011544168.1 | |
XM_011545868.1 | 550 | Missense Mutation | GAT,GCT | D,A 39 | XP_011544170.1 | |
XM_011545869.2 | 550 | UTR 5 | XP_011544171.1 | |||
XM_011545870.2 | 550 | UTR 5 | XP_011544172.1 | |||
XM_011545871.2 | 550 | UTR 5 | XP_011544173.1 | |||
XM_011545872.2 | 550 | UTR 5 | XP_011544174.1 | |||
XM_017023280.1 | 550 | Missense Mutation | GAT,GCT | D,A 45 | XP_016878769.1 | |
XM_017023281.1 | 550 | UTR 5 | XP_016878770.1 |