Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGTGATAACCCACAGAGCAGGT[C/T]GGGGTTTGGTGGGGGTGGTGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXW10 - F-box and WD repeat domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267585.1 | Intron | NP_001254514.1 | ||||
NM_001267586.1 | Intron | NP_001254515.1 | ||||
XM_011523625.1 | Intron | XP_011521927.1 | ||||
XM_011523626.2 | Intron | XP_011521928.1 | ||||
XM_017024020.1 | Intron | XP_016879509.1 | ||||
XM_017024021.1 | Intron | XP_016879510.1 |
TVP23B - trans-golgi network vesicle protein 23 homolog B (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |