Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTCGTTCTTTTTCGTCCTTTTCC[C/T]CGGTTGCTGCTTGCTGTGAGTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604182 MIM: 608855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MGC16275 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MGC16275 - uncharacterized protein MGC16275 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL38 - ribosomal protein L38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000999.3 | Intron | NP_000990.1 | ||||
NM_001035258.1 | Intron | NP_001030335.1 |
TTYH2 - tweety family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |