Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTCCAGCTCTGGGCAGCTCAC[C/T]TCATAGTCGTCTCTGCAGAGAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604313 MIM: 147557 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GALK1 - galactokinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000154.1 | 1020 | Silent Mutation | GAA,GAG | E,E 319 | NP_000145.1 |
ITGB4 - integrin subunit beta 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000213.4 | 1020 | Intron | NP_000204.3 | |||
NM_001005619.1 | 1020 | Intron | NP_001005619.1 | |||
NM_001005731.2 | 1020 | Intron | NP_001005731.1 | |||
NM_001321123.1 | 1020 | Intron | NP_001308052.1 | |||
XM_005257309.2 | 1020 | Intron | XP_005257366.1 | |||
XM_005257311.4 | 1020 | Intron | XP_005257368.1 | |||
XM_006721866.3 | 1020 | Intron | XP_006721929.1 | |||
XM_006721867.3 | 1020 | Intron | XP_006721930.1 | |||
XM_006721868.3 | 1020 | Intron | XP_006721931.1 | |||
XM_006721870.3 | 1020 | Intron | XP_006721933.1 | |||
XM_011524751.2 | 1020 | Intron | XP_011523053.1 | |||
XM_011524752.2 | 1020 | Intron | XP_011523054.1 |