Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGAGGAGGTCTCTGTTTGATGAG[C/T]GTCGTTTGCATCTGAAAGTTTCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604858 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRP68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRP68 - signal recognition particle 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260502.1 | 1647 | Missense Mutation | ACT,GCT | T,A 500 | NP_001247431.1 | |
NM_001260503.1 | 1647 | Missense Mutation | ACT,GCT | T,A 199 | NP_001247432.1 | |
NM_014230.3 | 1647 | Missense Mutation | ACT,GCT | T,A 538 | NP_055045.2 |