Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTAATGACACAGAAAAACAATA[C/T]TAAGTGAAACGTAACAATAAGCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPRS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COPRS - coordinator of PRMT5 and differentiation stimulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UTP6 - UTP6, small subunit processome component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018428.2 | Intron | NP_060898.2 | ||||
XM_005258012.2 | Intron | XP_005258069.1 | ||||
XM_011524997.2 | Intron | XP_011523299.1 | ||||
XM_017024845.1 | Intron | XP_016880334.1 |