Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTTGTCTCTCAGTACAATCAAG[A/G]TTAGGATCTGTAAGTTTCACTTATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1328 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1328 - KIAA1328 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322327.1 | Intron | NP_001309256.1 | ||||
NM_020776.1 | Intron | NP_065827.1 | ||||
XM_005258315.4 | Intron | XP_005258372.1 | ||||
XM_005258317.3 | Intron | XP_005258374.1 | ||||
XM_005258318.4 | Intron | XP_005258375.1 | ||||
XM_011526099.2 | Intron | XP_011524401.1 | ||||
XM_011526100.2 | Intron | XP_011524402.1 | ||||
XM_017025876.1 | Intron | XP_016881365.1 | ||||
XM_017025877.1 | Intron | XP_016881366.1 | ||||
XM_017025878.1 | Intron | XP_016881367.1 | ||||
XM_017025879.1 | Intron | XP_016881368.1 | ||||
XM_017025880.1 | Intron | XP_016881369.1 | ||||
XM_017025881.1 | Intron | XP_016881370.1 |