Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGCACTCCGGTGGCCCTGGGC[C/G]CCGTGACTAAGGCTCCTGTCAGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAF4B - TATA-box binding protein associated factor 4b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293725.1 | 181 | Missense Mutation | CCC,GCC | P,A 44 | NP_001280654.1 | |
NM_005640.2 | 181 | Missense Mutation | CCC,GCC | P,A 44 | NP_005631.1 | |
XM_005258339.3 | 181 | Missense Mutation | CCC,GCC | P,A 44 | XP_005258396.1 | |
XM_011526153.2 | 181 | Missense Mutation | CCC,GCC | P,A 44 | XP_011524455.1 | |
XM_017025932.1 | 181 | Missense Mutation | CCC,GCC | P,A 44 | XP_016881421.1 | |
XM_017025933.1 | 181 | Intron | XP_016881422.1 |