Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGCTGTAGCTCGTTCGCGAGGAC[C/T]ATCTCCATGCAAGCCATCCTTACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609966 MIM: 609293 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGN - gametogenetin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152657.3 | 1886 | Missense Mutation | AGT,GGT | S,G 547 | NP_689870.3 | |
XM_005258619.4 | 1886 | Missense Mutation | AGT,GGT | S,G 547 | XP_005258676.1 | |
XM_011526603.2 | 1886 | Missense Mutation | AGT,GGT | S,G 464 | XP_011524905.1 | |
XM_017026451.1 | 1886 | Missense Mutation | AGT,GGT | S,G 547 | XP_016881940.1 |
PSMD8 - proteasome 26S subunit, non-ATPase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRED3 - sprouty related EVH1 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |