Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGGCTATAAAATATCCCACATTC[C/T]TTACACTCAAAGGAGTTCTCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF780B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF780B - zinc finger protein 780B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005851.2 | 3654 | UTR 3 | NP_001005851.1 | |||
XM_005258590.4 | 3654 | UTR 3 | XP_005258647.1 | |||
XM_005258591.4 | 3654 | UTR 3 | XP_005258648.1 | |||
XM_005258592.3 | 3654 | UTR 3 | XP_005258649.1 | |||
XM_005258593.2 | 3654 | UTR 3 | XP_005258650.1 | |||
XM_005258595.3 | 3654 | UTR 3 | XP_005258652.1 | |||
XM_006723072.3 | 3654 | UTR 3 | XP_006723135.1 | |||
XM_011526593.2 | 3654 | UTR 3 | XP_011524895.1 | |||
XM_017026425.1 | 3654 | UTR 3 | XP_016881914.1 | |||
XM_017026426.1 | 3654 | UTR 3 | XP_016881915.1 | |||
XM_017026427.1 | 3654 | UTR 3 | XP_016881916.1 |