Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTAGTTATTTCTTTTATCAGAC[C/T]TCTCAGGTAGAAGCTTTGCCAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604086 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF155 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF155 - zinc finger protein 155 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260486.1 | Intron | NP_001247415.1 | ||||
NM_001260487.1 | Intron | NP_001247416.1 | ||||
NM_001260488.1 | Intron | NP_001247417.1 | ||||
NM_003445.3 | Intron | NP_003436.3 | ||||
NM_198089.2 | Intron | NP_932355.2 | ||||
XM_005259215.3 | Intron | XP_005259272.1 | ||||
XM_011527278.2 | Intron | XP_011525580.1 | ||||
XM_011527279.2 | Intron | XP_011525581.1 | ||||
XM_017027248.1 | Intron | XP_016882737.1 |
ZNF221 - zinc finger protein 221 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |