Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTAACTGAAGTGCTAACCTATT[G/T]ATCACACATCCAGGTGTTCCCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF229 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF229 - zinc finger protein 229 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278510.2 | 3149 | UTR 3 | NP_001265439.2 | |||
NM_014518.3 | 3149 | UTR 3 | NP_055333.3 | |||
XM_006723372.3 | 3149 | UTR 3 | XP_006723435.1 | |||
XM_011527292.1 | 3149 | UTR 3 | XP_011525594.1 | |||
XM_017027280.1 | 3149 | UTR 3 | XP_016882769.1 |