Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATTTACAGTCACAGAGACAGAGA[C/T]ACAAAGACACAACCCTGCACTGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608386 MIM: 605208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GFY - golgi associated olfactory signaling regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195256.1 | 2908 | Intron | NP_001182185.1 | |||
XM_011526299.2 | 2908 | Intron | XP_011524601.1 |
PTH2 - parathyroid hormone 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC17A7 - solute carrier family 17 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020309.3 | 2908 | UTR 3 | NP_064705.1 |