Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTCACCGCCGAGGAAGAGGAAG[A/G]CGCTCGTGGTCTGGCCCCGCAGCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130410 MIM: 606008 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDND2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDND2 - claudin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152353.2 | 1353 | Missense Mutation | GCC,GTC | A,V 98 | NP_689566.1 | |
XM_011526425.2 | 1353 | Missense Mutation | GCC,GTC | A,V 98 | XP_011524727.1 | |
XM_011526428.2 | 1353 | Missense Mutation | GCC,GTC | A,V 98 | XP_011524730.1 | |
XM_017026244.1 | 1353 | Silent Mutation | CGC,CGT | R,R 225 | XP_016881733.1 | |
XM_017026245.1 | 1353 | Silent Mutation | CGC,CGT | R,R 225 | XP_016881734.1 | |
XM_017026246.1 | 1353 | Missense Mutation | GCC,GTC | A,V 98 | XP_016881735.1 | |
XM_017026247.1 | 1353 | Silent Mutation | CGC,CGT | R,R 225 | XP_016881736.1 |
ETFB - electron transfer flavoprotein beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKG7 - natural killer cell granule protein 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |