Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGGGCTGAGCCAGGCTCTGTGA[C/T]CACCCAGGGGAGTCCCGTGACCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604810 MIM: 604812 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LILRA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LILRA1 - leukocyte immunoglobulin like receptor A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278318.1 | 164 | Intron | NP_001265247.1 | |||
NM_001278319.1 | 164 | Missense Mutation | ACC,ATC | T,I 39 | NP_001265248.1 | |
NM_006863.3 | 164 | Intron | NP_006854.1 |
LILRA2 - leukocyte immunoglobulin like receptor A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |