Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGTCTGTCATGGGTTGGCGCAC[C/G]CTGGGCCAGTGTTGTTGACCCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF549 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF549 - zinc finger protein 549 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199295.1 | Intron | NP_001186224.1 | ||||
NM_153263.2 | Intron | NP_694995.2 | ||||
XM_006723128.3 | Intron | XP_006723191.1 |
ZNF550 - zinc finger protein 550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |