Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGGCTGGGGGCCAGGGCTCTGGC[A/G]ACCTAGAGGTGTGGACGGCACAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114212 MIM: 603327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPS - calcyphosine | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004058.4 | 2992 | Intron | NP_004049.2 | |||
NM_080590.3 | 2992 | Intron | NP_542157.2 |
RANBP3 - RAN binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300865.1 | 2992 | UTR 3 | NP_001287794.1 | |||
NM_003624.2 | 2992 | UTR 3 | NP_003615.2 | |||
NM_007320.2 | 2992 | UTR 3 | NP_015559.2 | |||
NM_007322.2 | 2992 | UTR 3 | NP_015561.1 | |||
XM_006722928.1 | 2992 | UTR 3 | XP_006722991.1 | |||
XM_006722929.1 | 2992 | UTR 3 | XP_006722992.1 | |||
XM_011528391.1 | 2992 | UTR 3 | XP_011526693.1 | |||
XM_011528392.1 | 2992 | UTR 3 | XP_011526694.1 | |||
XM_011528393.1 | 2992 | UTR 3 | XP_011526695.1 | |||
XM_011528394.1 | 2992 | UTR 3 | XP_011526696.1 | |||
XM_017027405.1 | 2992 | UTR 3 | XP_016882894.1 | |||
XM_017027406.1 | 2992 | UTR 3 | XP_016882895.1 |
VMAC - vimentin-type intermediate filament associated coiled-coil protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |