Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGCCCAGCGGCTCACAGGCAGC[A/G]GTTTCTGTTGGTTGGGTCTGAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609458 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAN2B1 - mannosidase alpha class 2B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000528.3 | 1661 | Missense Mutation | CCG,CTG | P,L 669 | NP_000519.2 | |
NM_001173498.1 | 1661 | Missense Mutation | CCG,CTG | P,L 668 | NP_001166969.1 | |
XM_005259913.1 | 1661 | Missense Mutation | CCG,CTG | P,L 670 | XP_005259970.1 | |
XM_017026818.1 | 1661 | Missense Mutation | CCG,CTG | P,L 302 | XP_016882307.1 |