Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCGGTGGCCGGCGTAGAGCGC[A/G]GCCAGGGGCCGGAAGCGCGGACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609920 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDH22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDH22 - cadherin 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021248.2 | 3041 | Silent Mutation | GCC,GCT | A,A 813 | NP_067071.1 | |
XM_011528994.2 | 3041 | Silent Mutation | GCC,GCT | A,A 813 | XP_011527296.1 | |
XM_011528995.1 | 3041 | Silent Mutation | GCC,GCT | A,A 692 | XP_011527297.1 |