Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTGGAGGCAAGGATAGGATCCA[A/G]GTCTCCACTGAATCCTCAGCACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCM2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCM2L - CCM2 like scaffolding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080625.3 | Intron | NP_542192.2 | ||||
XM_006723706.3 | Intron | XP_006723769.1 | ||||
XM_011528566.1 | Intron | XP_011526868.1 | ||||
XM_011528567.2 | Intron | XP_011526869.1 | ||||
XM_011528568.2 | Intron | XP_011526870.1 | ||||
XM_011528569.1 | Intron | XP_011526871.1 |