Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGCGGACGCTATCTACGACCAC[A/G]TCAACGAGGGGGTGAGGGCCTGTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610843 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UQCR10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UQCR10 - ubiquinol-cytochrome c reductase, complex III subunit X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003684.1 | 169 | Missense Mutation | ATC,GTC | I,V 47 | NP_001003684.1 | |
NM_013387.3 | 169 | Missense Mutation | ATC,GTC | I,V 47 | NP_037519.2 |
ZMAT5 - zinc finger matrin-type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003692.1 | 169 | Intron | NP_001003692.1 | |||
NM_001318129.1 | 169 | Intron | NP_001305058.1 | |||
NM_019103.2 | 169 | Intron | NP_061976.1 |