Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGAAGTACTTAAAATTACCGTT[A/G]TACATGAACTTTGTGGACTGTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605928 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFIP1 - ADP ribosylation factor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025593.2 | 2608 | UTR 3 | NP_001020764.1 | |||
NM_001025595.2 | 2608 | UTR 3 | NP_001020766.1 | |||
NM_001287431.1 | 2608 | UTR 3 | NP_001274360.1 | |||
NM_001287432.1 | 2608 | UTR 3 | NP_001274361.1 | |||
NM_001287433.1 | 2608 | UTR 3 | NP_001274362.1 | |||
NM_014447.3 | 2608 | UTR 3 | NP_055262.1 | |||
XM_011531868.2 | 2608 | UTR 3 | XP_011530170.1 | |||
XM_011531869.2 | 2608 | UTR 3 | XP_011530171.1 | |||
XM_011531870.2 | 2608 | UTR 3 | XP_011530172.1 | |||
XM_011531871.2 | 2608 | UTR 3 | XP_011530173.1 | |||
XM_011531872.1 | 2608 | UTR 3 | XP_011530174.1 |
FHDC1 - FH2 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033393.2 | 2608 | Intron | NP_203751.2 | |||
XM_005263319.3 | 2608 | Intron | XP_005263376.1 | |||
XM_006714393.3 | 2608 | Intron | XP_006714456.1 | |||
XM_011532388.1 | 2608 | UTR 5 | XP_011530690.1 | |||
XM_011532389.1 | 2608 | Intron | XP_011530691.1 | |||
XM_017008787.1 | 2608 | Intron | XP_016864276.1 |