Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGTATCCTGGAAGGATCTAGAT[G/T]ATACCTTTCCCTGTCCTGTCTGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TRIM60 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TRIM60 - tripartite motif containing 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258025.1 | 309 | Missense Mutation | GAT,TAT | D,Y 49 | NP_001244954.1 | |
NM_152620.2 | 309 | Missense Mutation | GAT,TAT | D,Y 49 | NP_689833.1 | |
XM_011531683.2 | 309 | Missense Mutation | GAT,TAT | D,Y 49 | XP_011529985.1 |