Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCACCCTTATGGAAAGGTGGAGG[A/G]CATCTCCCTGATTTAATTAGCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
H6PD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
H6PD - hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282587.1 | 1331 | Intron | NP_001269516.1 | |||
NM_004285.3 | 1331 | Intron | NP_004276.2 | |||
XM_005263540.4 | 1331 | UTR 5 | XP_005263597.1 | |||
XM_006711052.3 | 1331 | Intron | XP_006711115.1 | |||
XM_017002865.1 | 1331 | Intron | XP_016858354.1 | |||
XM_017002866.1 | 1331 | Intron | XP_016858355.1 |