Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAGCGTGAGAATTAAAAAGGACC[C/T]TGAACTTGTGGTGACCGACCTGCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
AADACL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AADACL4 - arylacetamide deacetylase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013630.1 | 254 | Missense Mutation | CCT,CTT | P,L 85 | NP_001013652.1 | |
XM_017001152.1 | 254 | Missense Mutation | CCT,CTT | P,L 18 | XP_016856641.1 | |
XM_017001153.1 | 254 | Intron | XP_016856642.1 |