Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATTTCTAGAAACACACAAACAA[G/T]CCTCAAGAATAGAATAAAATAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611551 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBLN7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBLN7 - fibulin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128165.1 | Intron | NP_001121637.1 | ||||
NM_153214.2 | Intron | NP_694946.2 | ||||
XM_006712260.3 | Intron | XP_006712323.1 | ||||
XM_011510585.2 | Intron | XP_011508887.1 | ||||
XM_011510587.2 | Intron | XP_011508889.1 | ||||
XM_017003317.1 | Intron | XP_016858806.1 | ||||
XM_017003318.1 | Intron | XP_016858807.1 | ||||
XM_017003319.1 | Intron | XP_016858808.1 |
ZC3H8 - zinc finger CCCH-type containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032494.2 | Intron | NP_115883.2 | ||||
XM_011511998.1 | Intron | XP_011510300.1 | ||||
XM_017005107.1 | Intron | XP_016860596.1 |