Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTATAATATGAACCATATCTGTC[C/T]TCCTGAGTTCAGACGTGTTTTTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPOPL - speckle type BTB/POZ protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001664.2 | Intron | NP_001001664.1 | ||||
XM_005263655.4 | Intron | XP_005263712.1 | ||||
XM_005263656.4 | Intron | XP_005263713.1 | ||||
XM_005263657.4 | Intron | XP_005263714.1 |