Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGAGTATTGACATTTCAGCTACT[A/G]TCCCCAAGTTTACAAACACGCAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602332 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCAPH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCAPH - non-SMC condensin I complex subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281710.1 | 312 | Missense Mutation | ATC,GTC | I,V 88 | NP_001268639.1 | |
NM_001281711.1 | 312 | Missense Mutation | ATC,GTC | I,V 75 | NP_001268640.1 | |
NM_001281712.1 | 312 | UTR 5 | NP_001268641.1 | |||
NM_015341.4 | 312 | Missense Mutation | ATC,GTC | I,V 99 | NP_056156.2 | |
XM_005263908.3 | 312 | Missense Mutation | ATC,GTC | I,V 99 | XP_005263965.1 | |
XM_006712388.3 | 312 | Missense Mutation | ATC,GTC | I,V 99 | XP_006712451.1 |