Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGATGCTACACACAAGCTCCGCT[C/T]CTGGATTTTGATGAGGGCTGCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM228B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM228B - family with sequence similarity 228 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145710.1 | 454 | Intron | NP_001139182.1 | |||
NM_001291328.1 | 454 | Intron | NP_001278257.1 |
PFN4 - profilin family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199346.2 | 454 | Missense Mutation | NP_955378.1 | |||
XM_005264292.2 | 454 | Missense Mutation | XP_005264349.1 | |||
XM_006712007.3 | 454 | Missense Mutation | XP_006712070.1 | |||
XM_011532823.2 | 454 | Missense Mutation | XP_011531125.1 |