Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCACAAATCACACCAGGCACTG[A/T]ATGCGCCCATCCTGTCAGAAGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605158 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PXDN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PXDN - peroxidasin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012293.2 | 6040 | UTR 3 | NP_036425.1 | |||
XM_005264707.3 | 6040 | UTR 3 | XP_005264764.1 | |||
XM_011510396.1 | 6040 | UTR 3 | XP_011508698.1 | |||
XM_011510397.2 | 6040 | UTR 3 | XP_011508699.1 |