Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTTGGTGCCCACTTGAGACATT[A/C]AAAACAGTCACAACAGAACTATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC36 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC36 - coiled-coil domain containing 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135197.1 | 301 | Nonsense Mutation | TAA,TCA | *,S 68 | NP_001128669.1 | |
NM_178173.3 | 301 | Nonsense Mutation | TAA,TCA | *,S 68 | NP_835467.2 | |
XM_006713124.3 | 301 | Nonsense Mutation | TAA,TCA | *,S 68 | XP_006713187.1 | |
XM_011533670.2 | 301 | Nonsense Mutation | TAA,TCA | *,S 75 | XP_011531972.1 | |
XM_011533671.2 | 301 | Nonsense Mutation | TAA,TCA | *,S 75 | XP_011531973.1 | |
XM_011533672.2 | 301 | Nonsense Mutation | TAA,TCA | *,S 68 | XP_011531974.1 | |
XM_011533673.2 | 301 | Nonsense Mutation | TAA,TCA | *,S 68 | XP_011531975.1 |