Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACAGACGGTTCTTCCCCCAGTA[C/T]GTCAAGACCCTGGAGGGTGCAAGCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 128239 | |||||||||||||||||||||||
Literature Links: |
DAG1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
DAG1 - dystroglycan 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165928.3 | 365 | UTR 5 | NP_001159400.2 | |||
NM_001177634.2 | 365 | UTR 5 | NP_001171105.1 | |||
NM_001177635.2 | 365 | Intron | NP_001171106.1 | |||
NM_001177636.2 | 365 | UTR 5 | NP_001171107.1 | |||
NM_001177637.2 | 365 | Intron | NP_001171108.1 | |||
NM_001177638.2 | 365 | Intron | NP_001171109.1 | |||
NM_001177639.2 | 365 | Intron | NP_001171110.1 | |||
NM_001177640.2 | 365 | Intron | NP_001171111.1 | |||
NM_001177641.2 | 365 | Intron | NP_001171112.1 | |||
NM_001177642.2 | 365 | Intron | NP_001171113.1 | |||
NM_001177643.2 | 365 | Intron | NP_001171114.1 | |||
NM_001177644.2 | 365 | Intron | NP_001171115.1 | |||
NM_004393.5 | 365 | Intron | NP_004384.4 |