Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTAGTGACATGGAATTGAGGCCT[A/G]AATGACAAAAGTCAGTCATGTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPLX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPLX2 - complexin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008220.1 | Intron | NP_001008221.1 | ||||
NM_006650.3 | Intron | NP_006641.1 | ||||
XM_005265798.1 | Intron | XP_005265855.1 | ||||
XM_005265799.1 | Intron | XP_005265856.1 | ||||
XM_011534419.1 | Intron | XP_011532721.1 | ||||
XM_017008964.1 | Intron | XP_016864453.1 |