Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACCTGTCGTCACCTGGCTGCCA[C/T]CCTTATCTGAGGAAGCTCAGTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXD3 - exonuclease 3'-5' domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286823.1 | Intron | NP_001273752.1 | ||||
NM_017820.4 | Intron | NP_060290.3 | ||||
XM_011518810.1 | Intron | XP_011517112.1 | ||||
XM_011518814.1 | Intron | XP_011517116.1 | ||||
XM_011518815.1 | Intron | XP_011517117.1 | ||||
XM_017014861.1 | Intron | XP_016870350.1 | ||||
XM_017014862.1 | Intron | XP_016870351.1 | ||||
XM_017014863.1 | Intron | XP_016870352.1 | ||||
XM_017014864.1 | Intron | XP_016870353.1 | ||||
XM_017014865.1 | Intron | XP_016870354.1 | ||||
XM_017014866.1 | Intron | XP_016870355.1 | ||||
XM_017014867.1 | Intron | XP_016870356.1 |
NRARP - NOTCH-regulated ankyrin repeat protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |