Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATTAAAATGTCAAATGTCAACT[C/T]TAAATAGTCTATTGGGTGCTGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104615 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTUS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTUS2 - microtubule associated tumor suppressor candidate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A1 - solute carrier family 7 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003045.4 | 4241 | UTR 3 | NP_003036.1 | |||
XM_005266507.3 | 4241 | Intron | XP_005266564.1 | |||
XM_017020713.1 | 4241 | UTR 3 | XP_016876202.1 | |||
XM_017020714.1 | 4241 | UTR 3 | XP_016876203.1 | |||
XM_017020715.1 | 4241 | UTR 3 | XP_016876204.1 | |||
XM_017020716.1 | 4241 | Intron | XP_016876205.1 |