Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGCATATAAGGTCATTTTCTCA[C/T]CTGTGCTGCTCTGTTACACATTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602855 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMA6 - proteasome subunit alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282232.1 | Intron | NP_001269161.1 | ||||
NM_001282233.1 | Intron | NP_001269162.1 | ||||
NM_001282234.1 | Intron | NP_001269163.1 | ||||
NM_002791.2 | Intron | NP_002782.1 | ||||
XM_017021468.1 | Intron | XP_016876957.1 |