Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGTCTTGGCAGGTAAAGGGGCT[A/G]TTTTCATGACACATCTTGTTTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602329 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEL1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEL1L - SEL1L ERAD E3 ligase adaptor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244984.1 | 5814 | Intron | NP_001231913.1 | |||
NM_005065.5 | 5814 | UTR 3 | NP_005056.3 | |||
XM_005267988.2 | 5814 | Intron | XP_005268045.1 | |||
XM_005267989.3 | 5814 | Intron | XP_005268046.1 |