Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGCCAATCAGAAAACACAGAGG[A/G]GCCTTCCCTTGTGTGGGTCAGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605759 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB2 - ankyrin repeat and SOCS box containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM181A - family with sequence similarity 181 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207071.1 | Intron | NP_001194000.1 | ||||
NM_001207072.1 | Intron | NP_001194001.1 | ||||
NM_001207073.1 | Intron | NP_001194002.1 | ||||
NM_001207074.1 | Intron | NP_001194003.1 | ||||
NM_138344.4 | Intron | NP_612353.3 | ||||
XM_011537297.1 | Intron | XP_011535599.1 |
FAM181A-AS1 - FAM181A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |