Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGGTCTGTCCTTCAGCTAACT[A/G]TGCTGTTAAATTACTCTTCCTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601896 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAF3 - TNF receptor associated factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199427.1 | Intron | NP_001186356.1 | ||||
NM_003300.3 | Intron | NP_003291.2 | ||||
NM_145725.2 | Intron | NP_663777.1 | ||||
NM_145726.2 | Intron | NP_663778.1 | ||||
XM_011537116.2 | Intron | XP_011535418.1 | ||||
XM_011537117.2 | Intron | XP_011535419.1 | ||||
XM_011537118.2 | Intron | XP_011535420.1 | ||||
XM_017021617.1 | Intron | XP_016877106.1 | ||||
XM_017021618.1 | Intron | XP_016877107.1 | ||||
XM_017021619.1 | Intron | XP_016877108.1 | ||||
XM_017021620.1 | Intron | XP_016877109.1 |