Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTCCCATCCCATCACAGAACCTC[C/T]AGTTCCTGTCTGTCAATTGTGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102546294 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC102546294 - uncharacterized LOC102546294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPINK7 - serine peptidase inhibitor, Kazal type 7 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032566.2 | Intron | NP_115955.1 |
SPINK9 - serine peptidase inhibitor, Kazal type 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |