Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTAAGGGGAGAATCTGGCCTTA[C/T]CCTAGTGACTCCGAGGAGACTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNB3 - calcium voltage-gated channel auxiliary subunit beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000725.3 | Intron | NP_000716.2 | ||||
NM_001206915.1 | Intron | NP_001193844.1 | ||||
NM_001206916.1 | Intron | NP_001193845.1 | ||||
NM_001206917.1 | Intron | NP_001193846.1 | ||||
XM_011538727.2 | Intron | XP_011537029.1 | ||||
XM_011538728.2 | Intron | XP_011537030.1 | ||||
XM_017019957.1 | Intron | XP_016875446.1 | ||||
XM_017019958.1 | Intron | XP_016875447.1 | ||||
XM_017019959.1 | Intron | XP_016875448.1 |