Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTAGTGTCTGACCAGGGGACGA[C/T]AGTAACTTTTCTAAGGATTGAATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118950 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COQ10A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COQ10A - coenzyme Q10A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099337.1 | 1337 | UTR 3 | NP_001092807.1 | |||
NM_144576.3 | 1337 | UTR 3 | NP_653177.3 |
CS - citrate synthase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |